Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP70/HSPA1A, ac Lys77 Antibody, Novus Biologicals™
SDP

Catalog No. NBP261122 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP261122 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP261122 Supplier Novus Biologicals Supplier No. NBP261122
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HSP70/HSPA1A Polyclonal specifically detects HSP70/HSPA1A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen (Negative).

Specifications

Antigen HSP70/HSPA1A (ac Lys77)
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:1000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:60, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen (Negative) 1:50
Formulation PBS pH 7.4 and 50% glycerol with 0.09% Sodium Azide
Gene Alias dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B
Gene Symbols HSPA1A
Host Species Rabbit
Immunogen Synthetic peptide from the N-terminal of Human HSP70 Acetyl Lys77 (1-100 aa), conjugated to Keyhole Limpet Haemocyanin (KLH).
Purification Method Affinity Purified
Quantity 100 μL
Research Discipline Cancer, Cellular Markers, Chaperone Mediated Autophagy (CMA), Hypoxia, Membrane Trafficking and Chaperones, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3303
Test Specificity Binds to acetylated lysine 77 on HSP70. Detects 70 kDa band.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.