Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP70/HSPA1A, ac Lys77 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$610.00
Specifications
Antigen | HSP70/HSPA1A (ac Lys77) |
---|---|
Dilution | Western Blot 1:1000, ELISA 1:1000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:60, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen (Negative) 1:50 |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
HSP70/HSPA1A Polyclonal specifically detects HSP70/HSPA1A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen (Negative).Specifications
HSP70/HSPA1A (ac Lys77) | |
Polyclonal | |
Rabbit | |
dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B | |
HSPA1A | |
IgG | |
Affinity Purified |
Western Blot 1:1000, ELISA 1:1000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:60, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen (Negative) 1:50 | |
Unconjugated | |
Cancer, Cellular Markers, Chaperone Mediated Autophagy (CMA), Hypoxia, Membrane Trafficking and Chaperones, Phospho Specific | |
3303 | |
Synthetic peptide from the N-terminal of Human HSP70 Acetyl Lys77 (1-100 aa), conjugated to Keyhole Limpet Haemocyanin (KLH). | |
Primary | |
Binds to acetylated lysine 77 on HSP70. Detects 70 kDa band. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title