Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP70/HSPA1A Antibody (001), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB096705 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096705 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB096705 Supplier Novus Biologicals Supplier No. NBP289951V
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HSP70/HSPA1A Monoclonal specifically detects HSP70/HSPA1A in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen HSP70/HSPA1A
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1
Conjugate DyLight 405
Formulation 50 mM Sodium Borate
Gene Alias dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human HSP70/HSPA1A (P08107; Ala2-Asp641).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Chaperone Mediated Autophagy (CMA), Hypoxia, Membrane Trafficking and Chaperones, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3303
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.