Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP70/HSPA1A Antibody (1H11), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB259339488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259339488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB259339488 Supplier Novus Biologicals Supplier No. NBP259339AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HSP70/HSPA1A Monoclonal specifically detects HSP70/HSPA1A in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface).

Specifications

Antigen HSP70/HSPA1A
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1H11
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface)
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B
Gene Symbols HSPA1A
Host Species Mouse
Immunogen Human native HSP70 protein
Purification Method Protein G purified
Quantity 100 μL
Research Discipline Cancer, Cellular Markers, Chaperone Mediated Autophagy (CMA), Hypoxia, Membrane Trafficking and Chaperones, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3303
Test Specificity Binds the altered conformation of HSP70 embedded in the cell membrane and to intracellular HSP70 to a lesser extent.
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.