Learn More
Description
Specifications
Specifications
| Antigen | HSP70/HSPA1A |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 1H11 |
| Conjugate | HRP |
| Dilution | Western Blot |
| Formulation | PBS with No Preservative |
| Gene Alias | dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B |
| Gene Symbols | HSPA1A |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
