Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hsp70 interacting protein HIP Antibody (5C6), Biotin, Novus Biologicals™
SDP

Catalog No. NBP242671B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242671B 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242671B Supplier Novus Biologicals Supplier No. NBP242671B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hsp70 interacting protein HIP Monoclonal specifically detects Hsp70 interacting protein HIP in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CHST13
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 5C6
Conjugate Biotin
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias C4ST-3, C4ST3MGC119279, carbohydrate (chondroitin 4) sulfotransferase 13, carbohydrate sulfotransferase 13, Chondroitin 4-O-sulfotransferase 3, Chondroitin 4-sulfotransferase 3, EC 2.8.2.5, MGC119278, MGC119281
Gene Symbols CHST13
Host Species Mouse
Immunogen Recombinant human HIP/ST13 (1-369aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 166012
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.