Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP90 alpha Antibody, Novus Biologicals™
SDP

Catalog No. NB1202928 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1202928 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1202928 Supplier Novus Biologicals Supplier No. NB1202928
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 8 publications

HSP90 alpha Polyclonal specifically detects HSP90 alpha in Human, Mouse, Rat, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen HSP90 alpha
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 2 ug, Immunohistochemistry-Paraffin 5 ug/ml
Formulation PBS with 1mg/ml BSA with 0.05% Sodium Azide
Gene Accession No. P07901
Gene Alias FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38
Gene Symbols HSP90AA1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues P(2) E E T Q T Q D Q P M(12) of mouse HSP86.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Chaperone Mediated Autophagy (CMA)
Primary or Secondary Primary
Gene ID (Entrez) 3320
Test Specificity Detects Heat Shock Protein 86 (HSP 86). This does not detect HSP 84.
Target Species Human, Mouse, Rat, Primate, Rabbit, Sheep
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.