Learn More
Description
Specifications
Specifications
| Antigen | HSP90 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 2D12 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, Immunoprecipitation 1:10-1:500 |
| Formulation | PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide |
| Gene Alias | FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38 |
| Gene Symbols | HSP90AA1 |
| Host Species | Rat |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
