Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP90 Antibody (2D12), Novus Biologicals™
SDP

Catalog No. NBP19745005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
0.2 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP19745005 0.05 mL
NB018766 0.025 mL
NBP19745002 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP19745005 Supplier Novus Biologicals Supplier No. NBP1974500.05ML
Add to Cart
Add to Cart

Rat Monoclonal Antibody has been used in 4 publications

HSP90 Monoclonal specifically detects HSP90 in Human, Mouse, Rat, Bovine, Canine, Chicken, Fish, Fungi, Guinea Pig, Hamster, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen HSP90
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 2D12
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunoprecipitation 1:10-1:500
Formulation PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
Gene Alias FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38
Gene Symbols HSP90AA1
Host Species Rat
Immunogen Native mouse Hsp90.
Purification Method Ammonium sulfate precipitation
Quantity 0.05 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3320
Test Specificity Reacts with Beluga, and water mold
Target Species Human, Mouse, Rat, Bovine, Canine, Chicken, Fish, Fungi, Guinea Pig, Hamster, Primate, Rabbit, Sheep
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.