Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP90 Antibody (4B5), Novus Biologicals™
SDP

Catalog No. NB165484 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165484 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB165484 Supplier Novus Biologicals Supplier No. NBP315145
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

HSP90 Monoclonal antibody specifically detects HSP90 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation

Specifications

Antigen HSP90
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 4B5
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Formulation PBS (pH 7.4), 150mM NaCl, and 50% glycerol.
Gene Alias FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38
Host Species Rabbit
Immunogen A synthesized peptide derived from human HSP90. UniProt ID (P07900).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 3320
Target Species Human, Rat
Content And Storage Store at -20 to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.