Learn More
Description
Specifications
Specifications
| Antigen | HSP90AB1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | H9010 |
| Conjugate | APC |
| Dilution | Flow Cytometry |
| Gene Accession No. | P14625 |
| Gene Alias | D6S182, Heat shock 84 kDa, heat shock 90kD protein 1, beta, heat shock 90kDa protein 1, beta, Heat Shock Protein 90 Alpha Family Class B Member 1, Heat Shock Protein 90 KDa, heat shock protein 90kDa alpha (cytosolic), class B member 1, Heat Shock Protein 90kDa Alpha Family Class B Member 1, heat shock protein HSP 90-beta, HSP 84, HSP 90, HSP84, HSP90B, HSP90-BETA, HSPC2, HSPCB |
| Gene Symbols | HSP90AB1 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
