Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP90 beta Antibody (H9010), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB11061640I Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11061640I 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11061640I Supplier Novus Biologicals Supplier No. NB11061640IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HSP90 beta Monoclonal specifically detects HSP90 beta in Human, Mouse, Rat, Canine, Chicken, Fish, Hamster, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen HSP90AB1
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone H9010
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Accession No. P14625
Gene Alias D6S182, Heat shock 84 kDa, heat shock 90kD protein 1, beta, heat shock 90kDa protein 1, beta, Heat Shock Protein 90 Alpha Family Class B Member 1, Heat Shock Protein 90 KDa, heat shock protein 90kDa alpha (cytosolic), class B member 1, Heat Shock Protein 90kDa Alpha Family Class B Member 1, heat shock protein HSP 90-beta, HSP 84, HSP 90, HSP84, HSP90B, HSP90-BETA, HSPC2, HSPCB
Gene Symbols HSP90AB1
Host Species Mouse
Immunogen Recombinant human HSP90beta
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, ER Markers, Golgi Apparatus Markers, Hypoxia, Membrane Trafficking and Chaperones, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3326
Test Specificity Detects 90kDa. Detects HSP90 beta in all reactive species except in Chicken, where it detects both alpha and beta isoforms.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.