Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP90 beta Antibody (HSP90AB1/3952), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB389285 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB389285 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB389285 Supplier Novus Biologicals Supplier No. NBP308931JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HSP90 beta Monoclonal antibody specifically detects HSP90 beta in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen HSP90 beta
Applications Western Blot, Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone HSP90AB1/3952
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias Heat shock 84 kDa, heat shock 90kD protein 1, beta, heat shock 90kDa protein 1, beta, heat shock protein 90kDa alpha (cytosolic), class B member 1, heat shock protein beta, heat shock protein HSP 90-beta, HSP 84, HSP84, HSP90-BETA, HSP90BHSP 90, HSPC2D6S182, HSPCBFLJ26984
Host Species Mouse
Immunogen Recombinant human HSP90 beta protein fragment (around aa581-704) (exact sequence is proprietary) (Uniprot: P08238)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, ER Markers, Golgi Apparatus Markers, Hypoxia, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 3326
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.