Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPA12A Rabbit anti-Human, Mouse, Rat, Clone: 8Q8M3, Novus Biologicals™
SDP

Catalog No. NB166982 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166982 20 μg
NB166981 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166982 Supplier Novus Biologicals Supplier No. NBP31594320UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HSPA12A Monoclonal antibody specifically detects HSPA12A in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen HSPA12A
Applications Western Blot
Classification Monoclonal
Clone 8Q8M3
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias FLJ13874, heat shock 70 kDa protein 12A, heat shock 70kD protein 12A, heat shock 70kDa protein 12A, KIAA0417
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 50-150 of human HSPA12A (O43301).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259217
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.