Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSPA12A Rabbit anti-Human, Mouse, Rat, Clone: 8Q8M3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | HSPA12A |
---|---|
Clone | 8Q8M3 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
HSPA12A Monoclonal antibody specifically detects HSPA12A in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
HSPA12A | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
FLJ13874, heat shock 70 kDa protein 12A, heat shock 70kD protein 12A, heat shock 70kDa protein 12A, KIAA0417 | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human HSPA12A (O43301). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
8Q8M3 | |
Monoclonal | |
Purified | |
RUO | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
259217 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title