Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPA14 Rabbit anti-Human, Mouse, Monkey, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB093443 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB093443 20 μL
NB093442 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB093443 Supplier Novus Biologicals Supplier No. NBP30488120UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HSPA14 Polyclonal specifically detects HSPA14 in Human, Mouse, Monkey samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HSPA14
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias heat shock 70kDa protein 14, HSP70-4, HSP70L1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 240-509 of human HSPA14 (NP_057383.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51182
Target Species Human, Mouse, Monkey
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.