Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HspA1B Antibody, Novus Biologicals™
SDP

Catalog No. NBP216896 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP216896 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP216896 Supplier Novus Biologicals Supplier No. NBP216896
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HspA1B Polyclonal antibody specifically detects HspA1B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen HspA1B
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 1XPBS, 20% Glycerol (pH7). with 0.025% Proclin 300
Gene Accession No. P08107
Gene Alias FLJ54328, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1B, heat shock 70kDa protein 1B, HSP70.1/HSP70.2, HSP70-1/HSP70-2, HSP70-1B, HSP70-2, HSPA1, HSPA1A
Gene Symbols HSPA1B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Hsp70. The exact sequence is proprietary.
Molecular Weight of Antigen 70 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3304
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.