Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HspA1L Antibody, Novus Biologicals™
SDP

Catalog No. NBP133022 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP133022 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP133022 Supplier Novus Biologicals Supplier No. NBP133022
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HspA1L Polyclonal specifically detects HspA1L in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen HspA1L
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Alias heat shock 10kDa protein 1-like, Heat shock 70 kDa protein 1-Hom, Heat shock 70 kDa protein 1L, heat shock 70 kDa protein 1-like, heat shock 70kD protein-like 1, heat shock 70kDa protein 1-like, HSP70-1L, HSP70-HOM, HSP70T, hum70t
Gene Symbols HSPA1L
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human HSP70 1L. The exact sequence is proprietary.
Molecular Weight of Antigen 70 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3305
Test Specificity Tested in human. Not yet tested in any other species.
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.