Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSPA1L Mouse anti-Human, Mouse, Rat, Clone: 3E7H2, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | HSPA1L |
---|---|
Clone | 3E7H2 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7182
![]() ![]() |
Proteintech Group Inc
667801IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody well recognizes the endogenous HSPA1L protein in multiple cell lines.
In response to adverse changes in their environment, cells from many organisms increase the expression of a class of proteins referred to as heat shock proteins. One class of stress proteins, termed the "small heat shock proteins" is comprised of a diverse group of proteins from ∽15 to >30 kDa, with a single form found in yeast and human cells and multiple related forms found in higher plants. A direct relationship between elevated stress protein expression and neoplasia has been suggested from studies on estrogen-induced proteins in breast cancer cells. In addition, Hsp27 exhibits rapid increased phosphorylation in response to various mitogens, tumor promoter (e. g. phorbol esters) and calcium ionophores. The expression of a 24 kDa protein was shown to correlate with steroid hormone receptor in cell lines, organs, and tumors. The 24 kDa estrogen-induced protein was found to be expressed at high levels in carcinoma of the breast. Another study has suggested that levels of the Hsp27 correlate with endometrial adenocarcinomas. Interestingly, the regulation of human Hsp27 by estrogens and heat shock is reminiscent of the ecdysteroid and heat shock activation of the drosophila Hsp20 genes.Specifications
HSPA1L | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Heat shock 70 kDa protein 1L, HSP70 1L, HSP70 HOM, HSP70T, HSPA1L, hum70t | |
Hspa1l | |
IgG2b | |
Protein A | |
Antibody |
3E7H2 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P16627, P34931, P55063 | |
15482, 24963, 3305 | |
HSPA1L Fusion Protein Ag5182 | |
Primary | |
-20°C | |
HSPA1L |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title