Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPA2 Rabbit anti-Human, Mouse, Rat, Clone: S08-4A4, Novus Biologicals™
SDP

Catalog No. NB188244 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB188244 100 μg
NB188245 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB188244 Supplier Novus Biologicals Supplier No. NBP314944100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

HSPA2 Monoclonal antibody specifically detects HSPA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen HSPA2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone S08-4A4
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:20-1:100
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias Heat shock 70 kDa protein 2, heat shock 70kD protein 2, heat shock 70kDa protein 2, heat shock-related 70 kDa protein 2, HSP70-2, HSP70-3
Host Species Rabbit
Immunogen Recombinant protein of human HSPA2 (Uniprot#: P54652)
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 3306
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.