Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HspA4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP216898 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP216898 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP216898 Supplier Novus Biologicals Supplier No. NBP216898
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HspA4 Polyclonal specifically detects HspA4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen HspA4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. P34932
Gene Alias APG2, APG-2, heat shock 70 kDa protein 4, heat shock 70kD protein 4, heat shock 70kDa protein 4, Heat shock 70-related protein APG-2, heat shock protein, 110 kDa, HS24/P52, hsp70, hsp70RY, MGC131852, RY
Gene Symbols HSPA4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human HSPA4. The exact sequence is proprietary.
Molecular Weight of Antigen 94 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 3308
Test Specificity Human, Mouse. Expected to cross react based on sequence identity: Dog (96%), Rat (97%), Bovine (98%).
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.