Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HspA4L Antibody, Novus Biologicals™
SDP

Catalog No. NB10094907 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10094907 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10094907 Supplier Novus Biologicals Supplier No. NB10094907
Add to Cart
Add to Cart

Goat Polyclonal Antibody

HspA4L Polyclonal specifically detects HspA4L in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen HspA4L
Applications Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin, Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_055093.2
Gene Alias APG1, APG-1, heat shock 70 kDa protein 4L, heat shock 70 kDa protein 4-like protein, heat shock 70kDa protein 4-like, Heat shock 70-related protein APG-1, heat shock protein (hsp110 family), Osmotic stress protein 94, Osp94
Gene Symbols HSPA4L
Host Species Goat
Immunogen Peptide with sequence C-DSSQHTKSSGEMEVD corresponding to C-Terminus according to NP_055093.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22824
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.