Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HspA6 Antibody (165f), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP1974660.025MG

 View more versions of this product

Catalog No. NB018772


Only null left
Add to Cart

Description

Description

HspA6 Monoclonal specifically detects HspA6 in Human samples. It is validated for Western Blot.
Specifications

Specifications

HspA6
Monoclonal
Unconjugated
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
HSPA6
Synthetic peptide corresponding to the sequence near the C-terminus of human HspA6 Antibody (165f).
0.025 mg
Primary
Human
Purified
Western Blot
165f
Western Blot 1:1000
heat shock 70 kDa protein 6, Heat shock 70 kDa protein B', heat shock 70kD protein 6 (HSP70B'), heat shock 70kDa protein 6 (HSP70B'), HSP70B'
Mouse
Protein G purified
RUO
3310
Store at -20C. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only