Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HspA6 Antibody (165f), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$243.50 - $670.50

Specifications

Antigen HspA6
Clone 165f
Dilution Western Blot 1:1000
Applications Western Blot
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018772
SDP
View Documents
Novus Biologicals
NBP1974660.025MG
0.025 mg
Each for $243.50
Only null left
Add to Cart
 
NBP19746605
SDP
View Documents
Novus Biologicals
NBP1974660.05MG
0.05 mg
Each for $366.00
Only null left
Add to Cart
 
NBP19746602
SDP
View Documents
Novus Biologicals
NBP1974660.2MG
0.2 mg
Each for $670.50
Only null left
Add to Cart
 
Description

Description

HspA6 Monoclonal specifically detects HspA6 in Human samples. It is validated for Western Blot.
Specifications

Specifications

HspA6
Western Blot 1:1000
Monoclonal
Purified
RUO
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
3310
Synthetic peptide corresponding to the sequence near the C-terminus of human HspA6 Antibody (165f).
Primary
Store at -20C. Avoid freeze-thaw cycles.
165f
Western Blot
Unconjugated
Mouse
Human
heat shock 70 kDa protein 6, Heat shock 70 kDa protein B', heat shock 70kD protein 6 (HSP70B'), heat shock 70kDa protein 6 (HSP70B'), HSP70B'
HSPA6
IgG1
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.