Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HspA6 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB331297 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB331297 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB331297 Supplier Novus Biologicals Supplier No. NBP297536APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HspA6 Polyclonal antibody specifically detects HspA6 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen HspA6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias heat shock 70 kDa protein 6, Heat shock 70 kDa protein B', heat shock 70kD protein 6 (HSP70B'), heat shock 70kDa protein 6 (HSP70B'), HSP70B'
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human HspA6 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3310
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.