Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HspA6 Antibody (OTI1H3), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP27099A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27099A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27099A32 Supplier Novus Biologicals Supplier No. NBP270959AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HspA6 Monoclonal specifically detects HspA6 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HspA6
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1H3
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias heat shock 70 kDa protein 6, Heat shock 70 kDa protein B′, heat shock 70kD protein 6 (HSP70B′), heat shock 70kDa protein 6 (HSP70B′), HSP70B′
Gene Symbols HSPA6
Host Species Mouse
Immunogen Full length recombinant protein of human HspA6 (NP_002146) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3310.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.