Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPA8/HSC71/Hsc70 Antibody (13D3), Novus Biologicals™
SDP

Catalog No. NB1202788 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1202788 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1202788 Supplier Novus Biologicals Supplier No. NB1202788
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 19 publications

HSPA8/HSC71/Hsc70 Monoclonal specifically detects HSPA8/HSC71/Hsc70 in Human, Mouse, Rat, Bovine, Feline, Fish, Hamster, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Block/Neutralize.

Specifications

Antigen HSPA8/HSC71/Hsc70
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 13D3
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:5000, Simple Western reported by internal validation, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:20 - 1:200, Immunocytochemistry/Immunofluorescence 1:20 - 1:200, Immunoprecipitation 2 ul, Immunohistochemistry-Paraffin 1:20 - 1:200, Immunohistochemistry-Frozen 1:20 - 1:200, Immunoblotting, Block/Neutralize
Gene Accession No. P08107
Gene Alias Heat shock 70 kDa protein 8, heat shock 70kD protein 8, heat shock 70kDa protein 8, heat shock cognate 71 kDa protein, heat shock cognate protein 54, heat shock cognate protein, 71-kDa, HSC54, HSC70heat shock 70kd protein 10, HSC71, HSP71, HSP73constitutive heat shock protein 70, HSPA10, LAP1, lipopolysaccharide-associated protein 1, LPS-associated protein 1, MGC131511, MGC29929, NIP71, N-myristoyltransferase inhibitor protein 71
Gene Symbols HSPA8
Host Species Mouse
Immunogen Mouse spermatogenic cell protein.
Molecular Weight of Antigen 70 kDa
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Chaperone Mediated Autophagy (CMA)
Primary or Secondary Primary
Gene ID (Entrez) 3312
Test Specificity This displays slight cross-reactivity to HSP 70.
Target Species Human, Mouse, Rat, Bovine, Feline, Fish, Hamster, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.