Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPA8/HSC71/Hsc70 Antibody (1B5), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB362748 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362748 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362748 Supplier Novus Biologicals Supplier No. NBP197868DL594
Add to Cart
Add to Cart

Rat Monoclonal Antibody

HSPA8/HSC71/Hsc70 Monoclonal antibody specifically detects HSPA8/HSC71/Hsc70 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Guinea Pig, Hamster, Monkey, Rabbit, Sheep samples. It is validated for Western Blot, Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunomicroscopy

Specifications

Antigen HSPA8/HSC71/Hsc70
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunomicroscopy
Classification Monoclonal
Clone 1B5
Conjugate DyLight 594
Dilution Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Electron Microscopy
Formulation 50mM Sodium Borate
Gene Alias Heat shock 70 kDa protein 8, heat shock 70kD protein 8, heat shock 70kDa protein 8, heat shock cognate 71 kDa protein, heat shock cognate protein 54, heat shock cognate protein, 71-kDa, HSC54, HSC70heat shock 70kd protein 10, HSC71, HSP71, HSP73constitutive heat shock protein 70, HSPA10, LAP1, lipopolysaccharide-associated protein 1, LPS-associated protein 1, MGC131511, MGC29929, NIP71, N-myristoyltransferase inhibitor protein 71
Host Species Rat
Immunogen Native hamster Hsc70 (Hsp73).
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 3312
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Guinea Pig, Hamster, Monkey, Rabbit, Sheep
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.