Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPA8/HSC71/Hsc70 Antibody (1D9), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 3 publications

Supplier:  Novus Biologicals NBP1974980.2MG

 View more versions of this product

Catalog No. NBP19749802


Only null left
Add to Cart

Description

Description

HSPA8/HSC71/Hsc70 Monoclonal specifically detects HSPA8/HSC71/Hsc70 in Porcine, Plant samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

HSPA8/HSC71/Hsc70
Monoclonal
Unconjugated
PBS and 50% Glycerol with 0.09% Sodium Azide
HSPA8
Recombinant spinach Hsc70 (Hsp73).
0.2 mg
Primary
Pig, Plant
Purified
Western Blot, Immunoprecipitation
plant
Western Blot 1:1000, Immunoprecipitation 1:10-1:500
Heat shock 70 kDa protein 8, heat shock 70kD protein 8, heat shock 70kDa protein 8, heat shock cognate 71 kDa protein, heat shock cognate protein 54, heat shock cognate protein, 71-kDa, HSC54, HSC70heat shock 70kd protein 10, HSC71, HSP71, HSP73constitutive heat shock protein 70, HSPA10, LAP1, lipopolysaccharide-associated protein 1, LPS-associated protein 1, MGC131511, MGC29929, NIP71, N-myristoyltransferase inhibitor protein 71
Mouse
Protein G purified
RUO
3312
Store at -20C. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only