Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSPA8/HSC71/Hsc70 Antibody (1D9), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
$243.50 - $731.50
Specifications
Antigen | HSPA8/HSC71/Hsc70 |
---|---|
Clone | plant |
Dilution | Western Blot 1:1000, Immunoprecipitation 1:10-1:500 |
Applications | Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018790
![]() |
Novus Biologicals
NBP1974980.025MG |
0.025 mg |
Each for $243.50
|
|
|||||
NBP19749805
![]() |
Novus Biologicals
NBP1974980.05MG |
0.05 mg |
Each for $366.00
|
|
|||||
NBP19749802
![]() |
Novus Biologicals
NBP1974980.2MG |
0.2 mg |
Each for $731.50
|
|
|||||
Description
HSPA8/HSC71/Hsc70 Monoclonal specifically detects HSPA8/HSC71/Hsc70 in Porcine, Plant samples. It is validated for Western Blot, Immunoprecipitation.Specifications
HSPA8/HSC71/Hsc70 | |
Western Blot 1:1000, Immunoprecipitation 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
PBS and 50% Glycerol with 0.09% Sodium Azide | |
3312 | |
Recombinant spinach Hsc70 (Hsp73). | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
plant | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Pig, Plant | |
Heat shock 70 kDa protein 8, heat shock 70kD protein 8, heat shock 70kDa protein 8, heat shock cognate 71 kDa protein, heat shock cognate protein 54, heat shock cognate protein, 71-kDa, HSC54, HSC70heat shock 70kd protein 10, HSC71, HSP71, HSP73constitutive heat shock protein 70, HSPA10, LAP1, lipopolysaccharide-associated protein 1, LPS-associated protein 1, MGC131511, MGC29929, NIP71, N-myristoyltransferase inhibitor protein 71 | |
HSPA8 | |
IgG1 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title