Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPA8/HSC71/Hsc70 Antibody (5B7), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$203.51 - $739.91

Specifications

Antigen HSPA8/HSC71/Hsc70
Clone 5B7
Dilution Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018769
SDP
View Documents
Novus Biologicals
NBP1974540.025MG
0.025 mg
Each for $203.51
Only null left
Add to Cart
 
NBP19745405
SDP
View Documents
Novus Biologicals
NBP1974540.05MG
0.05 mg
Each for $394.97
Only null left
Add to Cart
 
NBP19745402
SDP
View Documents
Novus Biologicals
NBP1974540.2MG
0.2 mg
Each for $739.91
Only null left
Add to Cart
 
Description

Description

HSPA8/HSC71/Hsc70 Monoclonal specifically detects HSPA8/HSC71/Hsc70 in Plant samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

HSPA8/HSC71/Hsc70
Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500
Monoclonal
Purified
RUO
PBS and 50% Glycerol with 0.09% Sodium Azide
3312
Recombinant spinach Hsc70 (Hsp73).
Primary
Store at -20C. Avoid freeze-thaw cycles.
5B7
Immunoprecipitation, Western Blot
Unconjugated
Mouse
Plant
Heat shock 70 kDa protein 8, heat shock 70kD protein 8, heat shock 70kDa protein 8, heat shock cognate 71 kDa protein, heat shock cognate protein 54, heat shock cognate protein, 71-kDa, HSC54, HSC70heat shock 70kd protein 10, HSC71, HSP71, HSP73constitutive heat shock protein 70, HSPA10, LAP1, lipopolysaccharide-associated protein 1, LPS-associated protein 1, MGC131511, MGC29929, NIP71, N-myristoyltransferase inhibitor protein 71
HSPA8
IgG1
Protein G purified
Recognizes Plant HSC70.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.