Learn More
Description
Specifications
Specifications
| Antigen | HSPA8/HSC71/Hsc70 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 5B7 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500 |
| Formulation | PBS and 50% Glycerol with 0.09% Sodium Azide |
| Gene Alias | Heat shock 70 kDa protein 8, heat shock 70kD protein 8, heat shock 70kDa protein 8, heat shock cognate 71 kDa protein, heat shock cognate protein 54, heat shock cognate protein, 71-kDa, HSC54, HSC70heat shock 70kd protein 10, HSC71, HSP71, HSP73constitutive heat shock protein 70, HSPA10, LAP1, lipopolysaccharide-associated protein 1, LPS-associated protein 1, MGC131511, MGC29929, NIP71, N-myristoyltransferase inhibitor protein 71 |
| Gene Symbols | HSPA8 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
