Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HspB7 Antibody, Novus Biologicals™
SDP

Catalog No. NBP16907220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP16907220 20 μL
NBP169072 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP16907220 Supplier Novus Biologicals Supplier No. NBP16907220UL

Rabbit Polyclonal Antibody

HspB7 Polyclonal specifically detects HspB7 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen HspB7
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. P35385
Gene Alias Cardiovascular heat shock protein, CVHSP, DKFZp779D0968, FLJ32733, heat shock 27kDa protein family, member 7 (cardiovascular), heat shock protein beta-7, HspB7, member 7 (cardiovascular)
Gene Symbols HSPB7
Host Species Rabbit
Immunogen Synthetic peptides corresponding to Hspb7 (heat shock protein family, member 7 (cardiovascular)) The peptide sequence was selected from the middle region of Hspb7. Peptide sequence FGSFMLPHSEPLAFPARPGGQGNIKTLGDAYEFTVDMRDFSPEDIIVTTF.
Molecular Weight of Antigen 19 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27129
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.