Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HspBAP1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$480.74

Specifications

Antigen HspBAP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP180363
SDP
View Documents
Novus Biologicals
NBP180363
100 μL
Each for $480.74
Only null left
Add to Cart
 
NBP18036320
SDP
View Documents
Novus Biologicals
NBP18036320UL
20 μL N/A N/A N/A
Description

Description

HspBAP1 Polyclonal specifically detects HspBAP1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

HspBAP1
Polyclonal
Rabbit
NP_078886
79663
Synthetic peptide directed towards the N terminal of human HSPBAP1. Peptide sequence AAGSEATTPVIVAAGAGGEEGEHVKPFKPEKAKEIIMSLQQPAIFCNMVF.
Primary
Western Blot
Unconjugated
RUO
FLJ22623, heat shock 27 kDa associated protein, HSPB (heat shock 27kD) associated protein 1, HSPB (heat shock 27kDa) associated protein 1, HSPB1-associated protein 1,27 kDa heat shock protein-associated protein 1, PASS1FLJ39386, Protein associated with small stress protein 1, protein associating with small stress protein PASS1
HSPBAP1
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.