Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HspBAP1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP18036320 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP18036320 20 μL
NBP180363 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP18036320 Supplier Novus Biologicals Supplier No. NBP18036320UL

Rabbit Polyclonal Antibody

HspBAP1 Polyclonal specifically detects HspBAP1 in Human samples. It is validated for Western Blot.

Specifications

Antigen HspBAP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. NP_078886
Gene Alias FLJ22623, heat shock 27 kDa associated protein, HSPB (heat shock 27kD) associated protein 1, HSPB (heat shock 27kDa) associated protein 1, HSPB1-associated protein 1,27 kDa heat shock protein-associated protein 1, PASS1FLJ39386, Protein associated with small stress protein 1, protein associating with small stress protein PASS1
Gene Symbols HSPBAP1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal of human HSPBAP1. Peptide sequence AAGSEATTPVIVAAGAGGEEGEHVKPFKPEKAKEIIMSLQQPAIFCNMVF.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79663
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.