Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HspBP1 Antibody (OTI1D5), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB271761A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB271761A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB271761A48 Supplier Novus Biologicals Supplier No. NBP271761AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HspBP1 Monoclonal specifically detects HspBP1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen HspBP1
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI1D5
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FES1, Heat shock protein-binding protein 1, Hsp70 binding protein 1, hsp70 interacting protein, hsp70-binding protein 1, Hsp70-binding protein 2, Hsp70-interacting protein 1, Hsp70-interacting protein 2, HSPA (heat shock 70kDa) binding protein, cytoplasmic cochaperone 1, HSPBP, HspBP1, hspBP2
Gene Symbols HSPBP1
Host Species Mouse
Immunogen Full length recombinant protein of human HspBP1 (NP_036399) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23640.0
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.