Learn More
Description
Specifications
Specifications
| Antigen | HspBP1 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | OTI1D5 |
| Conjugate | Alexa Fluor 700 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | FES1, Heat shock protein-binding protein 1, Hsp70 binding protein 1, hsp70 interacting protein, hsp70-binding protein 1, Hsp70-binding protein 2, Hsp70-interacting protein 1, Hsp70-interacting protein 2, HSPA (heat shock 70kDa) binding protein, cytoplasmic cochaperone 1, HSPBP, HspBP1, hspBP2 |
| Gene Symbols | HSPBP1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
