Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPC014 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB095418 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB095418 20 μL
NB095417 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB095418 Supplier Novus Biologicals Supplier No. NBP30327420UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HSPC014 Polyclonal specifically detects HSPC014 in Human samples. It is validated for Western Blot.

Specifications

Antigen HSPC014
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias C13orf12, chromosome 13 open reading frame 12, HSPC014, hUMP1, PNAS-110, proteasome maturation protein, Proteassemblin, Protein UMP1 homolog, UMP12510048O06Rik, Voltage-gated K channel beta subunit 4.1, voltage-gated potassium channel beta subunit 4.1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-141 of human HSPC014 (NP_057016.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51371
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.