Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPH1/HSP105 Antibody (J1G12), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP104304A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP104304A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP104304A30 Supplier Novus Biologicals Supplier No. NBP104304AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HSPH1/HSP105 Monoclonal specifically detects HSPH1/HSP105 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HSPH1/HSP105
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone J1G12
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Antigen NY-CO-25, heat shock 105kD beta, heat shock 105kDa protein 1, heat shock 105kDa/110kDa protein 1, Heat shock 110 kDa protein, heat shock protein 105 kDa, HSP105A, HSP105B, HSP105heat shock 105kD alpha, HSP110, KIAA0201DKFZp686M05240, NY-CO-25
Gene Symbols HSPH1
Host Species Mouse
Immunogen Recombinant human HSP105 alpha (1-858 aa) purified from E. coli
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10808
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.