Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPH1/HSP105 Antibody (J1G12), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP104304IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP104304IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP104304IR Supplier Novus Biologicals Supplier No. NBP104304IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HSPH1/HSP105 Monoclonal specifically detects HSPH1/HSP105 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HSPH1/HSP105
Applications Western Blot, ELISA
Classification Monoclonal
Clone J1G12
Conjugate DyLight 755
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Antigen NY-CO-25, heat shock 105kD beta, heat shock 105kDa protein 1, heat shock 105kDa/110kDa protein 1, Heat shock 110 kDa protein, heat shock protein 105 kDa, HSP105A, HSP105B, HSP105heat shock 105kD alpha, HSP110, KIAA0201DKFZp686M05240, NY-CO-25
Gene Symbols HSPH1
Host Species Mouse
Immunogen Recombinant human HSP105 alpha (1-858 aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10808
Test Specificity Anti-human HSP105a mAb, clone J1G12, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human HSP105a protein.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.