Learn More
Description
Specifications
Specifications
| Antigen | HSPH1/HSP105 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | J1G12 |
| Conjugate | HRP |
| Dilution | Western Blot, ELISA |
| Formulation | PBS with No Preservative |
| Gene Alias | Antigen NY-CO-25, heat shock 105kD beta, heat shock 105kDa protein 1, heat shock 105kDa/110kDa protein 1, Heat shock 110 kDa protein, heat shock protein 105 kDa, HSP105A, HSP105B, HSP105heat shock 105kD alpha, HSP110, KIAA0201DKFZp686M05240, NY-CO-25 |
| Gene Symbols | HSPH1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
