Learn More
HSPH1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | HSPH1 |
|---|---|
| Concentration | 0.39 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8699
![]()
|
Proteintech Group Inc
133831AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody recognizes both HSP105alpha and HSP105beta isoforms. Western blot analysis using this antibody detected a major band around 100-110kD in Jurkat cells.
HSPH1 prevents the aggregation of denatured proteins in cells under severe stress, on which the ATP levels decrease markedly. This protein inhibits HSPA8/HSC70 ATPase and chaperone activities.Specifications
| HSPH1 | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q92598 | |
| 10808 | |
| HSPH1 Fusion Protein Ag4224 | |
| Primary | |
| -20°C | |
| HSPH1 |
| 0.39 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Antigen NY CO 25, Heat shock 110 kDa protein, Heat shock protein 105 kDa, HSP105, HSP105A, HSP105B, HSP110, HSPH1, KIAA0201, NY CO 25 | |
| HSPH1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.