Learn More
HSV1 Monoclonal Antibody (10A3), Zeta
Mouse Monoclonal Antibody Cocktail
$160.14
Specifications
Antigen | HSV1 |
---|---|
Clone | 10A3 |
Concentration | 200 μg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-221-5941
![]() |
ZETA CORPORATION
Z2215MS |
500 μL |
Each of 1 for $160.14
|
|
|||||
Description
A recommended positive control tissue for this product is Skin with HSV infection, however positive controls are not limited to this tissue type. The primary antibody is intended for laboratory professional use in the detection of the corresponding protein in formalin-fixed, paraffin-embedded tissue stained in manual qualitative immunohistochemistry (IHC) testing. This antibody is intended to be used after the primary diagnosis of tumor has been made by conventional histopathology using non-immunological histochemical stains.
HSV1 Monoclonal antibody specifically detects HSV1 in Human, Virus samples. It is validated for Immunohistochemistry (Paraffin)Specifications
HSV1 | |
200 μg/mL | |
Monoclonal | |
Liquid | |
RUO | |
tris with BSA, NP-40 and <0.1% sodium azide | |
Detergent-solubilized herpes simplex virus (HSV) type 1 infected cells | |
Primary | |
2-8°C |
10A3 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human, Virus | |
Herpes; Herpes Simplex; Herpes Simplex 1; Herpes Simplex I; herpes simplex virus 1; Herpes Simplex Virus Type 1; Herpes Virus; HHV; HSV; HSV Type 1; HSV1; KITH_HHV1S | |
IgG1 κ | |
Protein A | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.