Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HTF9C Antibody (OTI1C3), PerCP, Novus Biologicals™
SDP

Catalog No. NB271830PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB271830PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB271830PCP Supplier Novus Biologicals Supplier No. NBP271830PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HTF9C Monoclonal specifically detects HTF9C in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen HTF9C
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1C3
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias HTF9CHpaII tiny fragments locus 9c protein, MGC102728, TRM2 tRNA methyltransferase 2 homolog A (S. cerevisiae), tRNA (uracil-5-)-methyltransferase homolog A, tRNA methyltransferase 2 homolog A
Gene Symbols TRMT2A
Host Species Mouse
Immunogen Full length recombinant protein of human HTF9C (NP_073564) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 27037.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.