Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HTR12 Antibody, Novus Biologicals™
SDP

Catalog No. NBP118694 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP118694 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP118694 Supplier Novus Biologicals Supplier No. NBP118694
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

HTR12 Polyclonal specifically detects HTR12 in A. thaliana, Other, Plant samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HTR12
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:200-1:2000, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:500
Formulation Antiserum with No Preservative
Gene Accession No. P49450
Gene Alias CENH3, Centromeric histone CENH3, F6F3.17, F6F3_17, Histone H3 like centromeric protein HTR12, HTR 12
Gene Symbols HTR12
Host Species Rabbit
Immunogen Synthetic peptide derived from Nter domain of Arabidopsis thaliana (Mouse-ear cress) HTR12
Purification Method Unpurified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 839104
Test Specificity Reacts specifically with 17 kDa protein of HTR12
Reconstitution Reconstitute with 0.2 ml distilled water.
Target Species Other, Plant
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.