Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HTRA1/PRSS11 Antibody, Novus Biologicals™
SDP

Catalog No. NBP223869 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223869 0.1 mL
NBP223869SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP223869 Supplier Novus Biologicals Supplier No. NBP223869
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

HTRA1/PRSS11 Polyclonal specifically detects HTRA1/PRSS11 in Human, Mouse, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HTRA1/PRSS11
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:250-1: 1000, Simple Western 1:5000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Formulation 0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide
Gene Accession No. Q92743
Gene Alias ARMD7, EC 3.4.21, EC 3.4.21.108, High-temperature requirement A serine peptidase 1, HtrA serine peptidase 1, L56, ORF480, Serine protease 11, serine, 11 (IGF binding)
Gene Symbols HTRA1
Host Species Rabbit
Immunogen Within the range of amino acids 234-262of human HtrA1 were used as the immunogen, Gen Bank no NP_002766.1. This peptide sequence is in the serine protease domain of the HtrA1 protein.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5654
Test Specificity This antibody is reactive to Chimpanzee, Cow, Dog, Human, Mouse and Rat.
Target Species Human, Mouse, Rat, Bovine, Canine, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.