Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HTRA2/Omi Antibody (196C429), Novus Biologicals™
SDP

Catalog No. NB10056558 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056558 0.1 mg
NB10056558T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056558 Supplier Novus Biologicals Supplier No. NB10056558
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 3 publications

HTRA2/Omi Monoclonal specifically detects HTRA2/Omi in Human samples. It is validated for IF/IHC, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen HTRA2/Omi
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 196C429
Conjugate Unconjugated
Dilution Western Blot 2-3 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin reported in scientific literature (Lee et al (2003))
Gene Accession No. NP_713130.1
Gene Alias High temperature requirement protein A2, HtrA serine peptidase 2, HtrA2, HtrA-like serine protease, OMIOmi stress-regulated endoprotease, PARK13EC 3.4.21.108, PRSS25serine, 25, Serine protease 25, serine protease HTRA2, mitochondrial, Serine proteinase OMI
Gene Symbols HTRA2
Host Species Mouse
Immunogen This monoclonal antibody was developed by immunizing mice with a synthetic peptide containing amino acids 335-350 of human HtrA2.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Mitochondrial Mediated Pathway
Primary or Secondary Primary
Gene ID (Entrez) 27429
Test Specificity This antibody does not cross-react with mouse HtrA2.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.