Learn More
Description
Specifications
Specifications
| Antigen | HTRA2/Omi |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 196C429 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2-3 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin reported in scientific literature (Lee et al (2003)) |
| Gene Accession No. | NP_713130.1 |
| Gene Alias | High temperature requirement protein A2, HtrA serine peptidase 2, HtrA2, HtrA-like serine protease, OMIOmi stress-regulated endoprotease, PARK13EC 3.4.21.108, PRSS25serine, 25, Serine protease 25, serine protease HTRA2, mitochondrial, Serine proteinase OMI |
| Gene Symbols | HTRA2 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
