Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HTRA2/Omi Antibody (1B3), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP104325FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP104325FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP104325FR Supplier Novus Biologicals Supplier No. NBP104325FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HTRA2/Omi Monoclonal specifically detects HTRA2/Omi in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen HTRA2/Omi
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1B3
Conjugate DyLight 680
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias High temperature requirement protein A2, HtrA serine peptidase 2, HtrA2, HtrA-like serine protease, OMIOmi stress-regulated endoprotease, PARK13EC 3.4.21.108, PRSS25serine, 25, Serine protease 25, serine protease HTRA2, mitochondrial, Serine proteinase OMI
Gene Symbols HTRA2
Host Species Mouse
Immunogen Recombinant human HtrA2/Omi (134-458 aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Mitochondrial Mediated Pathway
Primary or Secondary Primary
Gene ID (Entrez) 27429
Test Specificity Anti-human HtrA2/Omi mAb, clone 1B3, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human HtrA2/Omi protein. Clone 1B3 is specific against human and mouse, but not rat.
Target Species Human, Mouse, Rat (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.