Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HuA/B/C/D Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | HuA/B/C/D |
---|---|
Concentration | 0.17 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8433
![]() ![]() |
Proteintech Group Inc
130321AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Embryonic lethal, abnormal vision, Drosophila-like 4 (ELAVL4) is a human homologue of the Drosophila gene ELAV, which is implicated in neuronal differentiation and maintece. The ELAVL proteins are mRNA binding proteins and all members of the family contain three RNA-recognition motifs. ELAVL-proteins also interact with inflammation-associated factors. The protein may be implicated in neurodegeneration through these complex mechanisms.Specifications
HuA/B/C/D | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat, Zebrafish | |
B5DF91, O09032, P26378, P70372, Q12926, Q14576, Q15717, Q60899, Q60900, Q61701, Q8CH84 | |
15568, 15569, 15571, 15572, 1993, 1994, 1995, 1996, 282824, 286973, 30732, 30736, 30737, 363854, 431719, 432358 | |
HuA/B/C/D Fusion Protein Ag3678 | |
Primary | |
-20°C | |
ELAVL1 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ELAV like protein 4, ELAVL4, Hu antigen D, Hu C/D, HUD, PNEM | |
Elavl1, elavl1a, Elavl2, Elavl3, Elavl4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title